Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOR1
Entrez Gene ID
9611
Gene Name
nuclear receptor corepressor 1
Gene Aliases
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location
Chr.17: 16030094 - 16216746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190440.1 NP_001177369.1 31 - 32 4859 74
NM_006311.3 NP_006302.2 32 - 33 5010 74
XM_005256866.4 XP_005256923.1 34 - 35 5245 74
XM_005256868.4 XP_005256925.1 34 - 35 5242 74
XM_005256871.4 XP_005256928.1 33 - 34 5218 74
XM_005256872.4 XP_005256929.1 33 - 34 5215 74
XM_005256873.4 XP_005256930.1 34 - 35 5197 74
XM_005256874.4 XP_005256931.1 33 - 34 5071 74
XM_006721601.3 XP_006721664.1 35 - 36 5377 74
XM_006721602.3 XP_006721665.1 35 - 36 5372 74
XM_006721603.3 XP_006721666.1 34 - 35 5245 74
XM_006721604.3 XP_006721667.1 32 - 33 5041 74
XM_011524083.2 XP_011522385.1 34 - 35 6156 74
XM_011524084.2 XP_011522386.1 34 - 35 5245 74
XM_011524085.2 XP_011522387.1 34 - 35 5245 74
XM_011524086.2 XP_011522388.1 32 - 33 5041 74
XM_017025396.1 XP_016880885.1 35 - 36 6122 74
XM_017025397.1 XP_016880886.1 35 - 36 5887 74
XM_017025398.1 XP_016880887.1 34 - 35 5329 74
XM_017025399.1 XP_016880888.1 34 - 35 5348 74
XM_017025400.1 XP_016880889.1 35 - 36 6119 74
XM_017025401.1 XP_016880890.1 35 - 36 5884 74
XM_017025402.1 XP_016880891.1 34 - 35 5168 74
XM_017025403.1 XP_016880892.1 34 - 35 5345 74
XM_017025404.1 XP_016880893.1 34 - 35 5836 74
XM_017025405.1 XP_016880894.1 33 - 34 5731 74
XM_017025406.1 XP_016880895.1 33 - 34 6129 74
XM_017025407.1 XP_016880896.1 33 - 34 5065 74
XM_017025408.1 XP_016880897.1 33 - 34 5137 74
XM_017025409.1 XP_016880898.1 34 - 35 5342 74
XM_017025410.1 XP_016880899.1 34 - 35 5838 74
XM_017025411.1 XP_016880900.1 33 - 34 5581 74
XM_017025412.1 XP_016880901.1 33 - 34 5358 74
XM_017025413.1 XP_016880902.1 33 - 34 5141 74
XM_017025414.1 XP_016880903.1 32 - 33 5048 74
XM_017025415.1 XP_016880904.1 33 - 34 5071 74
XM_017025416.1 XP_016880905.1 33 - 34 5679 74
XM_017025417.1 XP_016880906.1 33 - 34 5023 74
XM_017025418.1 XP_016880907.1 34 - 35 5245 74
XM_017025419.1 XP_016880908.1 33 - 34 5071 74
XM_017025420.1 XP_016880909.1 33 - 34 5023 74
GenBank mRNA AB028970.2 - 31 - 32 4833 74
AF044209.1 - 32 - 33 4981 74
AK295893.1 - 3 - 4 633 74
AK310405.1 - 8 - 9 1513 74
AL137641.1 - 2 - 3 563 74
BC167431.1 - 31 - 32 4834 74

View Full Product Details