Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NCOR1
- Entrez Gene ID
-
9611
- Gene Name
- nuclear receptor corepressor 1
- Gene Aliases
-
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
- Chromosome Location
-
Chr.17: 16030094 - 16216746 on Build GRCh38
View Full Product Details