Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NCOR1
Entrez Gene ID
9611
Gene Name
nuclear receptor corepressor 1
Gene Aliases
N-CoR, N-CoR1, PPP1R109, TRAC1, hN-CoR
Chromosome Location
Chr.17: 16030094 - 16216746 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190440.1 NP_001177369.1 42 - 43 6487 83
NM_006311.3 NP_006302.2 43 - 44 6995 83
XM_005256866.4 XP_005256923.1 45 - 46 7230 83
XM_005256868.4 XP_005256925.1 45 - 46 7227 83
XM_005256871.4 XP_005256928.1 44 - 45 7203 83
XM_005256872.4 XP_005256929.1 44 - 45 7200 83
XM_005256873.4 XP_005256930.1 45 - 46 7182 83
XM_005256874.4 XP_005256931.1 44 - 45 7056 83
XM_005256875.3 XP_005256932.1 44 - 45 7002 83
XM_006721601.3 XP_006721664.1 46 - 47 7362 83
XM_006721602.3 XP_006721665.1 46 - 47 7357 83
XM_006721603.3 XP_006721666.1 45 - 46 7191 83
XM_006721604.3 XP_006721667.1 43 - 44 7026 83
XM_011524083.2 XP_011522385.1 45 - 46 8141 83
XM_011524084.2 XP_011522386.1 45 - 46 7227 83
XM_011524085.2 XP_011522387.1 45 - 46 6873 83
XM_011524086.2 XP_011522388.1 43 - 44 6669 83
XM_017025396.1 XP_016880885.1 46 - 47 8107 83
XM_017025397.1 XP_016880886.1 46 - 47 7872 83
XM_017025398.1 XP_016880887.1 45 - 46 7314 83
XM_017025399.1 XP_016880888.1 45 - 46 7333 83
XM_017025400.1 XP_016880889.1 46 - 47 8104 83
XM_017025401.1 XP_016880890.1 46 - 47 7869 83
XM_017025402.1 XP_016880891.1 45 - 46 7153 83
XM_017025403.1 XP_016880892.1 45 - 46 7330 83
XM_017025404.1 XP_016880893.1 45 - 46 7821 83
XM_017025405.1 XP_016880894.1 44 - 45 7716 83
XM_017025406.1 XP_016880895.1 44 - 45 8114 83
XM_017025407.1 XP_016880896.1 44 - 45 7050 83
XM_017025408.1 XP_016880897.1 44 - 45 7122 83
XM_017025409.1 XP_016880898.1 45 - 46 7327 83
XM_017025410.1 XP_016880899.1 45 - 46 7823 83
XM_017025411.1 XP_016880900.1 44 - 45 7566 83
XM_017025412.1 XP_016880901.1 44 - 45 7343 83
XM_017025413.1 XP_016880902.1 44 - 45 7126 83
XM_017025414.1 XP_016880903.1 43 - 44 7033 83
XM_017025415.1 XP_016880904.1 44 - 45 7053 83
XM_017025416.1 XP_016880905.1 44 - 45 7664 83
XM_017025417.1 XP_016880906.1 44 - 45 7008 83
XM_017025418.1 XP_016880907.1 45 - 46 6870 83
XM_017025419.1 XP_016880908.1 44 - 45 6699 83
XM_017025420.1 XP_016880909.1 44 - 45 6651 83
GenBank mRNA AB028970.2 - 42 - 43 6461 83
AF044209.1 - 43 - 44 6966 83
AF087856.1 - 40 - 41 6522 83
AF303584.1 - 5 - 6 975 83
AF303585.1 - 12 - 13 2501 83
AK295893.1 - 14 - 15 2618 83
AK310405.1 - 19 - 20 3498 83
AL137641.1 - 13 - 14 2548 83
BC167431.1 - 42 - 43 6462 83

View Full Product Details