Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERMP1
Entrez Gene ID
79956
Gene Name
endoplasmic reticulum metallopeptidase 1
Gene Aliases
FXNA, KIAA1815, bA207C16.3
Chromosome Location
Chr.9: 5764061 - 5867091 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024896.2 NP_079172.2 3 - 4 817 117
XM_005251587.1 XP_005251644.1 3 - 4 788 117
XM_017015139.1 XP_016870628.1 4 - 5 610 117
GenBank mRNA AB058718.2 - 3 - 4 694 117
AK093217.1 - 2 - 3 184 117
AK127218.1 - 3 - 4 806 117
BC136771.1 - 3 - 4 791 117
BC136773.1 - 3 - 4 791 117
BC144404.1 - 3 - 4 791 117

View Full Product Details