Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CMA1
Entrez Gene ID
1215
Gene Name
chymase 1
Gene Aliases
CYH, MCT1, chymase
Chromosome Location
Chr.14: 24505355 - 24508265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001836.4 NP_001827.1 2 - 3 233 97
GenBank mRNA AB451464.1 - 2 - 3 203 97
BC069110.1 - 2 - 3 233 97
BC069370.1 - 2 - 3 233 97
BC069490.1 - 2 - 3 233 97
BC103975.1 - 2 - 3 97
DQ082727.1 - 2 - 3 184 97
M69136.1 - 2 - 3 218 97
S61334.1 - 1 - 2 140 97

View Full Product Details