The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000785.3 | NP_000776.1 | 6 - 7 | 1282 | 74 |
GenBank mRNA | AB005038.1 | - | 6 - 7 | 1251 | 74 |
AB005989.1 | - | 6 - 7 | 1130 | 74 | |
AF020192.1 | - | 6 - 7 | 1142 | 74 | |
AF246895.1 | - | 6 - 7 | 1130 | 74 | |
AK314953.1 | - | 6 - 7 | 1166 | 74 | |
BC136386.1 | - | 6 - 7 | 1160 | 74 | |
DQ925362.1 | - | 2 - 3 | 192 | 74 | |
DQ925363.1 | - | 1 - 2 | 89 | 74 | |
DQ925369.1 | - | 3 - 4 | 686 | 74 | |
DQ925370.1 | - | 4 - 5 | 498 | 74 | |
DQ925371.1 | - | 2 - 3 | 415 | 74 |