Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PWP1
Entrez Gene ID
11137
Gene Name
PWP1 homolog, endonuclein
Gene Aliases
IEF-SSP-9502
Chromosome Location
Chr.12: 107685732 - 107713162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317962.1 NP_001304891.1 10 - 11 994 118
NM_001317963.1 NP_001304892.1 10 - 11 1166 118
NM_007062.2 NP_008993.1 10 - 11 1138 118
GenBank mRNA AK223162.1 - 10 - 11 1062 118
AK290681.1 - 10 - 11 979 118
AK296252.1 - 10 - 11 995 118
AK297961.1 - 9 - 10 823 118
BC001652.2 - 10 - 11 1057 118
L07758.1 - 10 - 11 1057 118

View Full Product Details