Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PWP1
Entrez Gene ID
11137
Gene Name
PWP1 homolog, endonuclein
Gene Aliases
IEF-SSP-9502
Chromosome Location
Chr.12: 107685732 - 107713162 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317962.1 NP_001304891.1 9 - 10 930 69
NM_001317963.1 NP_001304892.1 9 - 10 1102 69
NM_007062.2 NP_008993.1 9 - 10 1074 69
GenBank mRNA AK223162.1 - 9 - 10 998 69
AK290681.1 - 9 - 10 915 69
AK296252.1 - 9 - 10 931 69
AK297961.1 - 8 - 9 759 69
BC001652.2 - 9 - 10 993 69
L07758.1 - 9 - 10 993 69

View Full Product Details