Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AGA
Entrez Gene ID
175
Gene Name
aspartylglucosaminidase
Gene Aliases
AGU, ASRG, GA
Chromosome Location
Chr.4: 177430774 - 177442503 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000027.3 NP_000018.2 6 - 7 822 82
GenBank mRNA AK312982.1 - 6 - 7 756 82
AW467768.1 - 2 - 3 82
BC012392.1 - 6 - 7 745 82
CR541715.1 - 6 - 7 694 82
M64073.1 - 6 - 7 697 82
X55762.1 - 6 - 7 721 82

View Full Product Details