Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RER1
Entrez Gene ID
11079
Gene Name
retention in endoplasmic reticulum sorting receptor 1
Gene Aliases
-
Chromosome Location
Chr.1: 2391314 - 2405444 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005244713.4 XP_005244770.2 1 - 2 1002 68

View Full Product Details