Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PASD1
Entrez Gene ID
139135
Gene Name
PAS domain containing 1
Gene Aliases
CT63, OXTES1
Chromosome Location
Chr.X: 151563535 - 151676739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173493.2 NP_775764.2 10 - 11 1169 120
XM_011531102.1 XP_011529404.1 10 - 11 1082 120
GenBank mRNA AY270020.1 - 10 - 11 975 120
BC040301.1 - 10 - 11 980 120

View Full Product Details