Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PASD1
Entrez Gene ID
139135
Gene Name
PAS domain containing 1
Gene Aliases
CT63, OXTES1
Chromosome Location
Chr.X: 151563535 - 151676739 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173493.2 NP_775764.2 4 - 5 540 88
XM_011531102.1 XP_011529404.1 4 - 5 453 88
GenBank mRNA AK097552.1 - 4 - 5 453 88
AY270020.1 - 4 - 5 346 88
BC040301.1 - 4 - 5 351 88

View Full Product Details