Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL4A2
Entrez Gene ID
1284
Gene Name
collagen type IV alpha 2
Gene Aliases
ICH, POREN2
Chromosome Location
Chr.13: 110307284 - 110513027 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CN397819.1 - 3 - 4 289 60

View Full Product Details