The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016614.2 | NP_057698.2 | 3 - 4 | 453 | 158 |
GenBank mRNA | AF201687.1 | - | 3 - 4 | 453 | 158 |
AF223469.1 | - | 3 - 4 | 444 | 158 | |
AJ269473.1 | - | 3 - 4 | 449 | 158 | |
AJ420495.1 | - | 2 - 3 | 654 | 158 | |
AK002168.1 | - | 3 - 4 | 428 | 158 | |
AK298699.1 | - | 3 - 4 | 658 | 158 | |
AK308836.1 | - | 3 - 4 | 456 | 158 | |
BC017553.2 | - | 3 - 4 | 465 | 158 | |
BC110375.1 | - | 3 - 4 | 434 | 158 |