Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCNG1
Entrez Gene ID
3755
Gene Name
potassium voltage-gated channel modifier subfamily G member 1
Gene Aliases
K13, KCNG, KV6.1, kH2
Chromosome Location
Chr.20: 51003656 - 51023176 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011528802.1 XP_011527104.1 1 - 2 259 89

View Full Product Details