Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FLNC
Entrez Gene ID
2318
Gene Name
filamin C
Gene Aliases
ABP-280, ABP280A, ABPA, ABPL, FLN2, MFM5, MPD4
Chromosome Location
Chr.7: 128830429 - 128859274 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127487.1 NP_001120959.1 24 - 25 4495 84
NM_001458.4 NP_001449.3 24 - 25 4495 84
GenBank mRNA AB208865.1 - 1 - 2 137 84
AB371585.1 - 24 - 25 4515 84
AF089841.1 - 24 - 25 4335 84
AF146692.1 - 24 - 25 84
AJ012737.1 - 24 - 25 4304 84

View Full Product Details