The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127487.1 | NP_001120959.1 | 2 - 3 | 811 | 86 |
NM_001458.4 | NP_001449.3 | 2 - 3 | 811 | 86 | |
GenBank mRNA | AB371585.1 | - | 2 - 3 | 831 | 86 |
AF146692.1 | - | 2 - 3 | 814 | 86 |