The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002202.2 | NP_002193.2 | 4 - 5 | 1318 | 58 |
XM_011543380.2 | XP_011541682.1 | 3 - 4 | 2436 | 58 | |
GenBank mRNA | AK290742.1 | - | 4 - 5 | 1036 | 58 |
BC031213.1 | - | 4 - 5 | 1319 | 58 | |
S70721.1 | - | 4 - 5 | 775 | 58 | |
U07559.1 | - | 4 - 5 | 1009 | 58 |