Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPIN2A
Entrez Gene ID
54466
Gene Name
spindlin family member 2A
Gene Aliases
DXF34, SPIN2, TDRD25, dJ323P24.1
Chromosome Location
Chr.X: 57134259 - 57138688 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA Y09858.1 - 1 - 1 85 67

View Full Product Details