Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GHITM
Entrez Gene ID
27069
Gene Name
growth hormone inducible transmembrane protein
Gene Aliases
DERP2, HSPC282, MICS1, My021, PTD010, TMBIM5
Chromosome Location
Chr.10: 84139429 - 84153555 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014394.2 NP_055209.2 4 - 5 549 117
GenBank mRNA AB009685.1 - 3 - 4 345 117
AF060923.1 - 4 - 5 423 117
AF078863.1 - 4 - 5 474 117
AF131820.1 - 4 - 5 476 117
AF161400.1 - 4 - 5 458 117
AK130094.1 - 4 - 5 452 117
AK130097.1 - 4 - 5 417 117
AK225318.1 - 4 - 5 426 117
AK292864.1 - 4 - 5 417 117
AK298332.1 - 4 - 5 431 117
AK299937.1 - 3 - 4 455 117
AY358824.1 - 4 - 5 424 117
BC010354.1 - 4 - 5 420 117
BC022872.1 - 4 - 5 117

View Full Product Details