The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286655.1 | NP_001273584.1 | 2 - 3 | 780 | 81 |
NM_001286656.1 | NP_001273585.1 | 2 - 3 | 364 | 81 | |
NM_023932.3 | NP_076421.2 | 2 - 3 | 364 | 81 | |
NM_206539.2 | NP_996262.1 | 2 - 3 | 780 | 81 | |
XM_005249308.4 | XP_005249365.1 | 1 - 2 | 1572 | 81 | |
XM_011514823.2 | XP_011513125.1 | 1 - 2 | 1026 | 81 | |
GenBank mRNA | AK055380.1 | - | 2 - 3 | 780 | 81 |
AY358126.1 | - | 2 - 3 | 230 | 81 | |
BC000230.1 | - | 2 - 3 | 196 | 81 | |
BC006425.1 | - | 2 - 3 | 196 | 81 | |
BC110320.1 | - | 2 - 3 | 332 | 81 | |
BI916137.1 | - | 2 - 3 | 288 | 81 | |
BM811564.1 | - | - | 329 | 81 | |
DA332624.1 | - | 2 - 3 | 231 | 81 |