The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000372.4 | NP_000363.1 | 4 - 5 | 1444 | 118 |
GenBank mRNA | AB775899.1 | - | 4 - 5 | 1362 | 118 |
AB775900.1 | - | 4 - 5 | 1362 | 118 | |
AB775901.1 | - | 4 - 5 | 1362 | 118 | |
J03581.1 | - | 4 - 5 | 1356 | 118 | |
M27160.1 | - | 4 - 5 | 1864 | 118 | |
M74314.1 | - | 4 - 5 | 1405 | 118 | |
U01873.1 | - | 4 - 5 | 1362 | 118 | |
Y00819.1 | - | 4 - 5 | 1368 | 118 |