The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042692.2 | NP_001036157.1 | 11 - 12 | 1836 | 90 |
NM_015910.5 | NP_056994.3 | 17 - 18 | 2651 | 90 | |
NR_122106.1 | 11 - 12 | 1832 | 90 | ||
XM_017004252.1 | XP_016859741.1 | 18 - 19 | 2475 | 90 | |
GenBank mRNA | AF131737.1 | - | 11 - 12 | 1836 | 90 |
AK001849.1 | - | 8 - 9 | 1447 | 90 | |
AK022647.1 | - | 11 - 12 | 1810 | 90 | |
AL833623.1 | - | 17 - 18 | 2437 | 90 | |
BC093752.1 | - | 11 - 12 | 1781 | 90 | |
BC093754.1 | - | 11 - 12 | 1781 | 90 | |
BX647132.1 | - | 12 - 13 | 5164 | 90 |