The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256550.1 | NP_001243479.1 | 1 - 2 | 234 | 111 |
NM_022474.3 | NP_071919.2 | 1 - 2 | 234 | 111 | |
GenBank mRNA | AK057160.1 | - | 1 - 2 | 94 | 111 |
AK098373.1 | - | 1 - 2 | 198 | 111 | |
AK291240.1 | - | 1 - 2 | 215 | 111 | |
AL832326.1 | - | 1 - 2 | 131 | 111 | |
AL832578.2 | - | 1 - 2 | 107 | 111 | |
BC053366.1 | - | 1 - 2 | 169 | 111 | |
BC095485.1 | - | 1 - 2 | 420 | 111 | |
BC129933.1 | - | 1 - 2 | 119 | 111 |