The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003367.3 | NP_003358.1 | 4 - 5 | 557 | 78 |
XM_005259197.3 | XP_005259254.1 | 4 - 5 | 456 | 78 | |
XM_011527260.2 | XP_011525562.1 | 4 - 5 | 456 | 78 | |
XM_011527261.2 | XP_011525563.1 | 4 - 5 | 456 | 78 | |
GenBank mRNA | AK297022.1 | - | 4 - 5 | 427 | 78 |
BC049821.1 | - | 4 - 5 | 470 | 78 | |
CR536504.1 | - | 4 - 5 | 433 | 78 | |
M77476.1 | - | 1 - 2 | 157 | 78 | |
X90824.1 | - | 2 - 3 | 296 | 78 | |
X90826.1 | - | 4 - 5 | 439 | 78 |