The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001270987.1 | NP_001257916.1 | 9 - 10 | 1165 | 107 |
NM_002342.2 | NP_002333.1 | 9 - 10 | 1360 | 107 | |
XM_005253688.1 | XP_005253745.1 | 8 - 9 | 1212 | 107 | |
GenBank mRNA | AK027080.1 | - | 9 - 10 | 1225 | 107 |
AK095418.1 | - | 10 - 11 | 1189 | 107 | |
AK128019.1 | - | 1 - 2 | 1419 | 107 | |
AK293187.1 | - | 9 - 10 | 1104 | 107 | |
BC026262.1 | - | 9 - 10 | 1232 | 107 | |
BX648074.1 | - | 1 - 2 | 1278 | 107 | |
L04270.1 | - | 9 - 10 | 1202 | 107 |