The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159995.2 | NP_001153467.1 | 7 - 8 | 771 | 79 |
NM_001159999.2 | NP_001153471.1 | 8 - 9 | 822 | 79 | |
NM_001160001.2 | NP_001153473.1 | 6 - 7 | 720 | 79 | |
NM_001322205.1 | NP_001309134.1 | 4 - 5 | 1863 | 79 | |
NM_001322206.1 | NP_001309135.1 | 4 - 5 | 1863 | 79 | |
NM_001322207.1 | NP_001309136.1 | 4 - 5 | 1863 | 79 | |
NM_013956.4 | NP_039250.2 | 8 - 9 | 1229 | 79 | |
XM_006716335.3 | XP_006716398.1 | 4 - 5 | 214 | 79 | |
XM_011544512.2 | XP_011542814.1 | 8 - 9 | 1012 | 79 | |
XM_017013367.1 | XP_016868856.1 | 6 - 7 | 722 | 79 | |
XM_017013368.1 | XP_016868857.1 | 6 - 7 | 1059 | 79 | |
XM_017013369.1 | XP_016868858.1 | 4 - 5 | 1831 | 79 | |
GenBank mRNA | AY207002.1 | - | 4 - 5 | 214 | 79 |
CN603661.1 | - | 4 - 5 | 500 | 79 | |
EF372273.1 | - | 6 - 7 | 556 | 79 | |
EF372274.1 | - | 6 - 7 | 556 | 79 | |
EF517296.1 | - | 8 - 9 | 658 | 79 | |
EF517297.1 | - | 7 - 8 | 607 | 79 | |
GQ983557.1 | - | 6 - 7 | 720 | 79 | |
M94166.1 | - | 8 - 9 | 804 | 79 | |
U02328.1 | - | 6 - 7 | 435 | 79 |