The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099222.1 | NP_001092692.1 | 13 - 14 | 1223 | 141 |
NM_001099223.1 | NP_001092693.1 | 13 - 14 | 1179 | 141 | |
NM_025103.2 | NP_079379.2 | 13 - 14 | 1215 | 141 | |
XM_011518035.2 | XP_011516337.1 | 13 - 14 | 1215 | 141 | |
XM_011518036.2 | XP_011516338.1 | 6 - 7 | 925 | 141 | |
XM_017015163.1 | XP_016870652.1 | 6 - 7 | 730 | 141 | |
XM_017015164.1 | XP_016870653.1 | 6 - 7 | 2171 | 141 | |
GenBank mRNA | AK023707.1 | - | 13 - 14 | 1176 | 141 |
AK026274.1 | - | 3 - 4 | 205 | 141 | |
AK225570.1 | - | 13 - 14 | 1176 | 141 | |
AY040325.1 | - | 13 - 14 | 1144 | 141 | |
BC023634.2 | - | 13 - 14 | 1160 | 141 | |
BC056875.1 | - | 11 - 12 | 923 | 141 | |
BC113524.1 | - | 12 - 13 | 988 | 141 | |
BC113526.1 | - | 12 - 13 | 989 | 141 |