The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099222.1 | NP_001092692.1 | 16 - 17 | 1497 | 124 |
NM_001099223.1 | NP_001092693.1 | 16 - 17 | 1453 | 124 | |
NM_025103.2 | NP_079379.2 | 16 - 17 | 1489 | 124 | |
XM_011518035.2 | XP_011516337.1 | 16 - 17 | 1489 | 124 | |
XM_011518036.2 | XP_011516338.1 | 9 - 10 | 1199 | 124 | |
XM_017015163.1 | XP_016870652.1 | 9 - 10 | 1004 | 124 | |
XM_017015164.1 | XP_016870653.1 | 9 - 10 | 2445 | 124 | |
GenBank mRNA | AK023707.1 | - | 16 - 17 | 1450 | 124 |
AK026274.1 | - | 6 - 7 | 479 | 124 | |
AK225570.1 | - | 16 - 17 | 1450 | 124 | |
AY040325.1 | - | 16 - 17 | 1418 | 124 | |
BC023634.2 | - | 16 - 17 | 1434 | 124 | |
BC056875.1 | - | 14 - 15 | 1197 | 124 | |
BC113524.1 | - | 15 - 16 | 1262 | 124 | |
BC113526.1 | - | 15 - 16 | 1263 | 124 |