The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099222.1 | NP_001092692.1 | 6 - 7 | 637 | 130 |
NM_001099223.1 | NP_001092693.1 | 6 - 7 | 593 | 130 | |
NM_001099224.1 | NP_001092694.1 | 6 - 7 | 629 | 130 | |
NM_025103.2 | NP_079379.2 | 6 - 7 | 629 | 130 | |
XM_011518035.2 | XP_011516337.1 | 6 - 7 | 629 | 130 | |
GenBank mRNA | AK225570.1 | - | 6 - 7 | 590 | 130 |
AY040325.1 | - | 6 - 7 | 558 | 130 | |
BC023634.2 | - | 6 - 7 | 574 | 130 | |
BC056875.1 | - | 4 - 5 | 337 | 130 | |
BC107742.1 | - | 6 - 7 | 527 | 130 | |
BC113524.1 | - | 5 - 6 | 402 | 130 | |
BC113526.1 | - | 5 - 6 | 403 | 130 |