The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142807.1 | NP_001136279.1 | 15 - 16 | 1592 | 68 |
XM_011511404.2 | XP_011509706.1 | 16 - 17 | 1716 | 68 | |
XM_011511405.2 | XP_011509707.1 | 16 - 17 | 1734 | 68 | |
XM_011511406.2 | XP_011509708.1 | 16 - 17 | 1707 | 68 | |
XM_011511407.2 | XP_011509709.1 | 15 - 16 | 1467 | 68 | |
XM_011511409.1 | XP_011509711.1 | 16 - 17 | 1716 | 68 | |
XM_011511411.2 | XP_011509713.1 | 15 - 16 | 1599 | 68 | |
XM_011511413.1 | XP_011509715.1 | 16 - 17 | 1716 | 68 | |
XM_011511415.2 | XP_011509717.1 | 16 - 17 | 1716 | 68 | |
XM_011511416.2 | XP_011509718.1 | 16 - 17 | 1716 | 68 | |
XM_011511419.2 | XP_011509721.1 | 15 - 16 | 1626 | 68 | |
XM_011511432.2 | XP_011509734.1 | 16 - 17 | 1544 | 68 | |
XM_011511434.2 | XP_011509736.1 | 7 - 8 | 849 | 68 | |
XM_017004433.1 | XP_016859922.1 | 8 - 9 | 791 | 68 | |
XM_017004434.1 | XP_016859923.1 | 8 - 9 | 751 | 68 | |
GenBank mRNA | AK001904.1 | - | 10 - 11 | 975 | 68 |
AK300513.1 | - | 15 - 16 | 1591 | 68 | |
BC131514.1 | - | 9 - 10 | 882 | 68 |