Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACOXL
Entrez Gene ID
55289
Gene Name
acyl-CoA oxidase-like
Gene Aliases
-
Chromosome Location
Chr.2: 110732539 - 111118571 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142807.1 NP_001136279.1 8 - 9 844 82
XM_011511404.2 XP_011509706.1 8 - 9 878 82
XM_011511405.2 XP_011509707.1 8 - 9 896 82
XM_011511406.2 XP_011509708.1 8 - 9 869 82
XM_011511407.2 XP_011509709.1 7 - 8 629 82
XM_011511409.1 XP_011509711.1 8 - 9 878 82
XM_011511411.2 XP_011509713.1 8 - 9 878 82
XM_011511413.1 XP_011509715.1 8 - 9 878 82
XM_011511414.2 XP_011509716.1 8 - 9 878 82
XM_011511415.2 XP_011509717.1 8 - 9 878 82
XM_011511416.2 XP_011509718.1 8 - 9 878 82
XM_011511419.2 XP_011509721.1 8 - 9 878 82
XM_011511420.2 XP_011509722.1 8 - 9 878 82
XM_011511421.2 XP_011509723.1 8 - 9 878 82
XM_011511422.1 XP_011509724.1 8 - 9 878 82
XM_011511423.1 XP_011509725.1 8 - 9 878 82
XM_011511427.2 XP_011509729.1 8 - 9 878 82
XM_011511428.2 XP_011509730.1 8 - 9 878 82
XM_011511429.2 XP_011509731.1 8 - 9 878 82
XM_011511432.2 XP_011509734.1 8 - 9 706 82
XM_017004431.1 XP_016859920.1 8 - 9 878 82
XM_017004432.1 XP_016859921.1 8 - 9 878 82
GenBank mRNA AK001904.1 - 2 - 3 137 82
AK300513.1 - 8 - 9 843 82
BC131514.1 - 2 - 3 134 82

View Full Product Details