Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- MAP2
- Entrez Gene ID
-
4133
- Gene Name
- microtubule associated protein 2
- Gene Aliases
-
MAP2A, MAP2B, MAP2C
- Chromosome Location
-
Chr.2: 209424047 - 209734118 on Build GRCh38
View Full Product Details