Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL13A1
Entrez Gene ID
1305
Gene Name
collagen type XIII alpha 1 chain
Gene Aliases
CMS19, COLXIIIA1
Chromosome Location
Chr.10: 69801885 - 69959148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130103.1 NP_001123575.1 18 - 19 1534 68
NM_001320951.1 NP_001307880.1 18 - 19 1504 68
NM_080798.3 NP_542988.3 14 - 15 1363 68
NM_080800.3 NP_542990.3 17 - 18 1477 68
NM_080801.3 NP_542991.3 17 - 18 1468 68
NM_080802.3 NP_542992.3 17 - 18 1468 68
NM_080805.3 NP_542995.3 15 - 16 1381 68
XM_011539292.2 XP_011537594.1 18 - 19 1507 68
XM_011539293.2 XP_011537595.1 18 - 19 1507 68
XM_011539294.2 XP_011537596.1 18 - 19 1507 68
XM_011539295.2 XP_011537597.1 18 - 19 1507 68
XM_017015676.1 XP_016871165.1 18 - 19 1507 68
XM_017015677.1 XP_016871166.1 18 - 19 1507 68
XM_017015678.1 XP_016871167.1 17 - 18 1480 68
XM_017015679.1 XP_016871168.1 18 - 19 1507 68
XM_017015680.1 XP_016871169.1 18 - 19 1507 68
XM_017015681.1 XP_016871170.1 18 - 19 1507 68
XM_017015682.1 XP_016871171.1 18 - 19 1507 68
XM_017015683.1 XP_016871172.1 17 - 18 1471 68
XM_017015684.1 XP_016871173.1 17 - 18 1471 68
XM_017015685.1 XP_016871174.1 18 - 19 1507 68
XM_017015686.1 XP_016871175.1 18 - 19 1507 68
XM_017015687.1 XP_016871176.1 18 - 19 1507 68
XM_017015688.1 XP_016871177.1 18 - 19 1507 68
XM_017015689.1 XP_016871178.1 18 - 19 1507 68
XM_017015690.1 XP_016871179.1 15 - 16 1417 68
XM_017015691.1 XP_016871180.1 18 - 19 1507 68
XM_017015692.1 XP_016871181.1 17 - 18 1471 68
XM_017015693.1 XP_016871182.1 18 - 19 1507 68
XM_017015694.1 XP_016871183.1 14 - 15 1366 68
XM_017015695.1 XP_016871184.1 14 - 15 1366 68
XM_017015696.1 XP_016871185.1 15 - 16 660 68
XM_017015697.1 XP_016871186.1 17 - 18 938 68
GenBank mRNA AJ293624.1 - 18 - 19 1495 68
BC136385.1 - 14 - 15 1133 68
CN297094.1 - 5 - 6 244 68
M59217.1 - 17 - 18 804 68

View Full Product Details