Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- COL13A1
- Entrez Gene ID
-
1305
- Gene Name
- collagen type XIII alpha 1 chain
- Gene Aliases
-
CMS19, COLXIIIA1
- Chromosome Location
-
Chr.10: 69801885 - 69959148 on Build GRCh38
View Full Product Details