Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL13A1
Entrez Gene ID
1305
Gene Name
collagen type XIII alpha 1 chain
Gene Aliases
CMS19, COLXIIIA1
Chromosome Location
Chr.10: 69801885 - 69959148 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130103.1 NP_001123575.1 39 - 40 2683 68
NM_001320951.1 NP_001307880.1 37 - 38 2572 68
NM_080798.3 NP_542988.3 34 - 35 2467 68
NM_080800.3 NP_542990.3 37 - 38 2590 68
NM_080801.3 NP_542991.3 38 - 39 2617 68
NM_080802.3 NP_542992.3 36 - 37 2536 68
NM_080805.3 NP_542995.3 33 - 34 2362 68
XM_011539292.2 XP_011537594.1 40 - 41 2719 68
XM_011539293.2 XP_011537595.1 39 - 40 2674 68
XM_011539294.2 XP_011537596.1 39 - 40 2656 68
XM_011539295.2 XP_011537597.1 38 - 39 2638 68
XM_017015676.1 XP_016871165.1 40 - 41 2710 68
XM_017015677.1 XP_016871166.1 39 - 40 2674 68
XM_017015678.1 XP_016871167.1 38 - 39 2629 68
XM_017015679.1 XP_016871168.1 38 - 39 2629 68
XM_017015680.1 XP_016871169.1 38 - 39 2623 68
XM_017015681.1 XP_016871170.1 38 - 39 2611 68
XM_017015682.1 XP_016871171.1 37 - 38 2587 68
XM_017015683.1 XP_016871172.1 37 - 38 2587 68
XM_017015684.1 XP_016871173.1 37 - 38 2575 68
XM_017015685.1 XP_016871174.1 37 - 38 2569 68
XM_017015686.1 XP_016871175.1 37 - 38 2560 68
XM_017015687.1 XP_016871176.1 37 - 38 2551 68
XM_017015688.1 XP_016871177.1 36 - 37 2533 68
XM_017015689.1 XP_016871178.1 37 - 38 2524 68
XM_017015690.1 XP_016871179.1 34 - 35 2506 68
XM_017015691.1 XP_016871180.1 36 - 37 2488 68
XM_017015692.1 XP_016871181.1 35 - 36 2488 68
XM_017015693.1 XP_016871182.1 36 - 37 2482 68
XM_017015694.1 XP_016871183.1 33 - 34 2419 68
XM_017015695.1 XP_016871184.1 32 - 33 2341 68
XM_017015696.1 XP_016871185.1 36 - 37 1809 68
XM_017015697.1 XP_016871186.1 36 - 37 1997 68
GenBank mRNA AJ293624.1 - 39 - 40 2644 68
AK124476.1 - 1 - 2 2139 68
BC136385.1 - 34 - 35 2237 68
BC139913.1 - 10 - 11 1235 68
BY796799.2 - 9 - 10 484 68

View Full Product Details