The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022909.3 | NP_075060.1 | 7 - 8 | 568 | 150 |
GenBank mRNA | AB035124.1 | - | 7 - 8 | 481 | 150 |
AK123761.1 | - | 6 - 7 | 486 | 150 | |
AK290746.1 | - | 7 - 8 | 551 | 150 | |
BC012024.1 | - | 7 - 8 | 544 | 150 | |
BC015355.1 | - | 7 - 8 | 550 | 150 | |
KU178758.1 | - | 7 - 8 | 481 | 150 | |
KU178759.1 | - | 7 - 8 | 440 | 150 |