Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HEBP1
Entrez Gene ID
50865
Gene Name
heme binding protein 1
Gene Aliases
HBP, HEBP
Chromosome Location
Chr.12: 12974864 - 13000309 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015987.4 NP_057071.2 2 - 3 419 79
GenBank mRNA AF117615.1 - 2 - 3 302 79
AF167473.1 - 2 - 3 224 79
AK289877.1 - 2 - 3 383 79
BC016277.2 - 2 - 3 254 79
BI550915.1 - 2 - 3 423 79
BT007294.1 - 2 - 3 224 79
CF126276.1 - 2 - 3 420 79

View Full Product Details