The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300754.1 | NP_001287683.1 | 8 - 9 | 1240 | 63 |
NM_001300756.1 | NP_001287685.1 | 8 - 9 | 1606 | 63 | |
NM_178833.5 | NP_849155.2 | 9 - 10 | 1777 | 63 | |
XM_006714085.2 | XP_006714148.1 | 9 - 10 | 1776 | 63 | |
XM_006714086.3 | XP_006714149.1 | 8 - 9 | 1605 | 63 | |
GenBank mRNA | AK172823.1 | - | 9 - 10 | 1777 | 63 |
AK297010.1 | - | 8 - 9 | 1151 | 63 | |
AK299888.1 | - | 8 - 9 | 1165 | 63 | |
BC009732.1 | - | 6 - 7 | 845 | 63 | |
BC047447.1 | - | 9 - 10 | 1307 | 63 |