Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THAP6
Entrez Gene ID
152815
Gene Name
THAP domain containing 6
Gene Aliases
-
Chromosome Location
Chr.4: 75513951 - 75547334 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK309187.1 - 3 - 4 354 145

View Full Product Details