The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001048205.1 | NP_001041670.1 | 15 - 16 | 1774 | 123 |
NM_005132.2 | NP_005123.2 | 16 - 17 | 1673 | 123 | |
XM_011537421.1 | XP_011535723.1 | 15 - 16 | 1946 | 123 | |
XM_017021841.1 | XP_016877330.1 | 15 - 16 | 1820 | 123 | |
GenBank mRNA | AF006264.1 | - | 15 - 16 | 1246 | 123 |
AK001435.1 | - | 16 - 17 | 1673 | 123 | |
AK291191.1 | - | 16 - 17 | 1805 | 123 | |
AL832973.1 | - | 13 - 14 | 2163 | 123 | |
BC004159.2 | - | 15 - 16 | 1330 | 123 | |
BC010887.1 | - | 15 - 16 | 1707 | 123 | |
BC019326.1 | - | 9 - 10 | 692 | 123 | |
CR457263.1 | - | 15 - 16 | 1246 | 123 |