The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001242644.1 | NP_001229573.1 | 4 - 5 | 582 | 98 |
NM_002865.2 | NP_002856.1 | 5 - 6 | 654 | 98 | |
GenBank mRNA | AF070629.1 | - | 1 - 2 | 75 | 98 |
AF498930.1 | - | 5 - 6 | 356 | 98 | |
AK297576.1 | - | 4 - 5 | 556 | 98 | |
AK312344.1 | - | 5 - 6 | 577 | 98 | |
BC008929.2 | - | 5 - 6 | 544 | 98 | |
BT019695.1 | - | 5 - 6 | 356 | 98 | |
M28213.1 | - | 5 - 6 | 396 | 98 | |
X12953.1 | - | 5 - 6 | 564 | 98 |