The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006716335.3 | XP_006716398.1 | 2 - 3 | 140 | 60 |
GenBank mRNA | AY207002.1 | - | 2 - 3 | 140 | 60 |
U02327.1 | - | 5 - 6 | 342 | 60 |