The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271998.1 | NP_001258927.1 | 2 - 3 | 376 | 130 |
NR_073545.1 | 4 - 5 | 572 | 130 | ||
GenBank mRNA | BI828974.1 | - | 4 - 5 | 456 | 130 |
BX354553.2 | - | 2 - 3 | 305 | 130 | |
KC407675.1 | - | 4 - 5 | 294 | 130 |