The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204831.1 | NP_001191760.1 | 7 - 8 | 1392 | 85 |
NM_001204832.1 | NP_001191761.1 | 6 - 7 | 1194 | 85 | |
NM_001204833.1 | NP_001191762.1 | 5 - 6 | 1059 | 85 | |
NM_001204834.1 | NP_001191763.1 | 6 - 7 | 822 | 85 | |
NM_018075.3 | NP_060545.3 | 7 - 8 | 1392 | 85 | |
XM_011533885.2 | XP_011532187.2 | 7 - 8 | 1358 | 85 | |
XM_011533887.2 | XP_011532189.1 | 7 - 8 | 1358 | 85 | |
XM_011533889.2 | XP_011532191.1 | 7 - 8 | 1358 | 85 | |
XM_011533890.2 | XP_011532192.1 | 7 - 8 | 1358 | 85 | |
XM_017006716.1 | XP_016862205.1 | 7 - 8 | 1358 | 85 | |
XM_017006717.1 | XP_016862206.1 | 7 - 8 | 1310 | 85 | |
XM_017006718.1 | XP_016862207.1 | 7 - 8 | 1337 | 85 | |
XM_017006719.1 | XP_016862208.1 | 6 - 7 | 1112 | 85 | |
XM_017006720.1 | XP_016862209.1 | 7 - 8 | 1337 | 85 | |
XM_017006721.1 | XP_016862210.1 | 6 - 7 | 1121 | 85 | |
XM_017006722.1 | XP_016862211.1 | 6 - 7 | 1094 | 85 | |
GenBank mRNA | AK096302.1 | - | 7 - 8 | 1392 | 85 |
AK131223.1 | - | 6 - 7 | 1110 | 85 | |
AK292368.1 | - | 7 - 8 | 1277 | 85 | |
AK295969.1 | - | 5 - 6 | 994 | 85 | |
AK297949.1 | - | 7 - 8 | 1335 | 85 | |
AL832508.1 | - | 7 - 8 | 1259 | 85 | |
BC038855.1 | - | 6 - 7 | 720 | 85 |