The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139498.1 | NP_001132970.1 | 5 - 6 | 733 | 76 |
NM_001139500.1 | NP_001132972.1 | 6 - 7 | 901 | 76 | |
NM_001139501.1 | NP_001132973.1 | 7 - 8 | 1011 | 76 | |
NM_001139502.1 | NP_001132974.1 | 5 - 6 | 654 | 76 | |
NM_004114.3 | NP_004105.1 | 4 - 5 | 1266 | 76 | |
NM_033642.2 | NP_378668.1 | 4 - 5 | 529 | 76 | |
XM_005262399.1 | XP_005262456.1 | 6 - 7 | 1104 | 76 | |
GenBank mRNA | AF100143.1 | - | 4 - 5 | 834 | 76 |
AF100144.1 | - | 4 - 5 | 529 | 76 | |
AK289960.1 | - | 4 - 5 | 944 | 76 | |
AK297545.1 | - | 6 - 7 | 932 | 76 | |
AK303685.1 | - | 5 - 6 | 733 | 76 | |
AK316170.1 | - | 5 - 6 | 810 | 76 | |
BC012347.1 | - | 4 - 5 | 1263 | 76 | |
BC034340.1 | - | 4 - 5 | 970 | 76 | |
BU195176.1 | - | 5 - 6 | 670 | 76 | |
U66198.1 | - | 4 - 5 | 604 | 76 |