The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001042517.1 | NP_001035982.1 | 24 - 25 | 3253 | 156 |
NM_001258366.1 | NP_001245295.1 | 23 - 24 | 3220 | 156 | |
NM_001258367.1 | NP_001245296.1 | 22 - 23 | 3115 | 156 | |
NM_001258368.1 | NP_001245297.1 | 22 - 23 | 3043 | 156 | |
NM_001258369.1 | NP_001245298.1 | 24 - 25 | 3253 | 156 | |
NM_030932.3 | NP_112194.2 | 18 - 19 | 2424 | 156 | |
XM_006719876.1 | XP_006719939.1 | 18 - 19 | 2404 | 156 | |
XM_011535258.2 | XP_011533560.1 | 24 - 25 | 3196 | 156 | |
GenBank mRNA | AB244756.1 | - | 23 - 24 | 3001 | 156 |
AB244757.1 | - | 22 - 23 | 2896 | 156 | |
AB244758.1 | - | 22 - 23 | 2824 | 156 | |
AY750055.1 | - | 24 - 25 | 3126 | 156 | |
AY818645.1 | - | 24 - 25 | 3034 | 156 | |
BC034952.1 | - | 18 - 19 | 2428 | 156 | |
BC048963.1 | - | 18 - 19 | 2380 | 156 | |
BC068504.1 | - | 18 - 19 | 2380 | 156 |