The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256685.1 | NP_001243614.1 | 4 - 5 | 449 | 109 |
NM_001256689.1 | NP_001243618.1 | 3 - 4 | 363 | 109 | |
NM_001256693.1 | NP_001243622.1 | 2 - 3 | 267 | 109 | |
NM_014791.3 | NP_055606.1 | 4 - 5 | 449 | 109 | |
NR_046337.1 | 3 - 4 | 363 | 109 | ||
XM_011518076.2 | XP_011516378.1 | 4 - 5 | 628 | 109 | |
XM_011518077.1 | XP_011516379.1 | 5 - 6 | 524 | 109 | |
XM_011518078.2 | XP_011516380.1 | 5 - 6 | 711 | 109 | |
XM_011518079.1 | XP_011516381.1 | 4 - 5 | 401 | 109 | |
XM_011518080.1 | XP_011516382.1 | 4 - 5 | 416 | 109 | |
XM_011518081.2 | XP_011516383.1 | 4 - 5 | 434 | 109 | |
XM_011518082.2 | XP_011516384.1 | 3 - 4 | 542 | 109 | |
XM_011518083.2 | XP_011516385.1 | 4 - 5 | 625 | 109 | |
XM_011518084.2 | XP_011516386.1 | 3 - 4 | 237 | 109 | |
XM_011518085.1 | XP_011516387.1 | 2 - 3 | 233 | 109 | |
XM_011518086.2 | XP_011516388.1 | 4 - 5 | 416 | 109 | |
GenBank mRNA | AB183427.1 | - | 3 - 4 | 314 | 109 |
AK222811.1 | - | 4 - 5 | 448 | 109 | |
AK296820.1 | - | 3 - 4 | 362 | 109 | |
AK299164.1 | - | 4 - 5 | 401 | 109 | |
AK300821.1 | - | 3 - 4 | 317 | 109 | |
AK301131.1 | - | 2 - 3 | 199 | 109 | |
AK309470.1 | - | 2 - 3 | 224 | 109 | |
BC014039.2 | - | 4 - 5 | 403 | 109 | |
BP365611.1 | - | 4 - 5 | 451 | 109 | |
D79997.1 | - | 4 - 5 | 435 | 109 |