The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001300753.1 | NP_001287682.1 | 7 - 8 | 1443 | 135 |
NM_001300755.1 | NP_001287684.1 | 7 - 8 | 1541 | 135 | |
NM_001300757.1 | NP_001287686.1 | 6 - 7 | 1196 | 135 | |
NM_001300758.1 | NP_001287687.1 | 6 - 7 | 1192 | 135 | |
NM_144567.4 | NP_653168.2 | 7 - 8 | 1518 | 135 | |
NR_125333.1 | 5 - 6 | 1065 | 135 | ||
XM_005273344.1 | XP_005273401.1 | 7 - 8 | 1539 | 135 | |
XM_005273345.1 | XP_005273402.1 | 7 - 8 | 1585 | 135 | |
XM_005273346.2 | XP_005273403.1 | 7 - 8 | 1338 | 135 | |
XM_005273347.3 | XP_005273404.1 | 6 - 7 | 1210 | 135 | |
XM_017002774.1 | XP_016858263.1 | 7 - 8 | 1467 | 135 | |
XM_017002775.1 | XP_016858264.1 | 8 - 9 | 1766 | 135 | |
XM_017002776.1 | XP_016858265.1 | 8 - 9 | 1790 | 135 | |
XM_017002777.1 | XP_016858266.1 | 8 - 9 | 1836 | 135 | |
XM_017002778.1 | XP_016858267.1 | 8 - 9 | 1579 | 135 | |
GenBank mRNA | AF510741.1 | - | 6 - 7 | 1147 | 135 |
AK093947.1 | - | 2 - 3 | 1942 | 135 | |
AK294479.1 | - | 7 - 8 | 1317 | 135 | |
AK295116.1 | - | 7 - 8 | 1447 | 135 | |
AK299365.1 | - | 5 - 6 | 975 | 135 | |
AK310946.1 | - | 5 - 6 | 684 | 135 | |
AL079275.1 | - | 7 - 8 | 1476 | 135 | |
BC047469.1 | - | 6 - 7 | 1108 | 135 |