Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANGEL2
Entrez Gene ID
90806
Gene Name
angel homolog 2
Gene Aliases
Ccr4d, KIAA0759L
Chromosome Location
Chr.1: 212992182 - 213015875 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144567.4 NP_653168.2 2 - 3 592 148
XM_005273344.1 XP_005273401.1 2 - 3 613 148
XM_005273345.1 XP_005273402.1 2 - 3 659 148
XM_005273346.2 XP_005273403.1 2 - 3 412 148
XM_017002774.1 XP_016858263.1 2 - 3 541 148
GenBank mRNA AK295116.1 - 2 - 3 521 148
AL079275.1 - 2 - 3 547 148
AY468376.1 - 1 - 2 322 148
BC016966.1 - 1 - 2 336 148

View Full Product Details